Assay Details
| Gene Symbol: Gpbp1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029868.2 | NP_084144.2 | 11 - 12 | 2437 | 66 | |
| GenBank mRNA | AK037224.1 | - | 11 - 12 | 2458 | 66 | |
| AK077148.1 | - | 11 - 12 | 2407 | 66 | ||
| AK133761.1 | - | 11 - 12 | 2453 | 66 | ||
| BC031428.1 | - | 10 - 11 | 1510 | 4021073 | 66 | |
| BC066034.1 | - | 11 - 12 | 2437 | 6836605 | 66 | |
More Information
Set Membership: |
|

Gpbp1l1
Genomic Map