Assay Details
| Gene Symbol: Pik3r3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181585.5 | NP_853616.1 | 4 - 5 | 886 | 131 | |
| GenBank mRNA | AK031936.1 | - | 4 - 5 | 803 | 131 | |
| AK132341.1 | - | 4 - 5 | 1161 | 131 | ||
| AK135693.1 | - | 4 - 5 | 1195 | 131 | ||
| AK161822.1 | - | 4 - 5 | 1154 | 131 | ||
| AK163413.1 | - | 4 - 5 | 1160 | 131 | ||
| BC053102.1 | - | 4 - 5 | 886 | 6405534 | 131 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pik3r3
Genomic Map