Assay Details
| Gene Symbol: Sec63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153055.3 | NP_694695.3 | 12 - 13 | 1474 | 128 | |
| GenBank mRNA | AK036629.1 | - | 12 - 13 | 1165 | 128 | |
| AK049737.1 | - | 10 - 11 | 3595 | 128 | ||
| AY024346.1 | - | 12 - 13 | 1271 | 128 | ||
| AY238936.1 | - | 12 - 13 | 1244 | 128 | ||
| BC019366.1 | - | 12 - 13 | 1238 | 5066908 | 128 | |
| BC029774.1 | - | 4 - 5 | 478 | 128 | ||
| BC031846.1 | - | 12 - 13 | 1238 | 5066822 | 128 | |
| BC059816.1 | - | 12 - 13 | 1473 | 128 | ||
| BC080810.1 | - | 11 - 12 | 2777 | 128 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sec63
Genomic Map