Assay Details
| Gene Symbol: Sec63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153055.3 | NP_694695.3 | 8 - 9 | 1005 | 147 | |
| GenBank mRNA | AK036629.1 | - | 8 - 9 | 700 | 147 | |
| AK049737.1 | - | 6 - 7 | 3126 | 147 | ||
| AY024346.1 | - | 8 - 9 | 802 | 147 | ||
| AY238936.1 | - | 8 - 9 | 775 | 147 | ||
| BC019366.1 | - | 8 - 9 | 769 | 5066908 | 147 | |
| BC031846.1 | - | 8 - 9 | 769 | 5066822 | 147 | |
| BC059816.1 | - | 8 - 9 | 1004 | 147 | ||
| BC080810.1 | - | 7 - 8 | 2308 | 147 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sec63
Genomic Map