Assay Details
| Gene Symbol: Ddx56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026538.3 | NP_080814.1 | 1 - 2 | 69 | 62 | |
| GenBank mRNA | AK011136.1 | - | 1 - 2 | 79 | 62 | |
| AK076047.1 | - | 1 - 2 | 87 | 62 | ||
| AK076975.1 | - | 1 - 2 | 69 | 62 | ||
| AK077514.1 | - | 1 - 2 | 82 | 62 | ||
| AK145628.1 | - | 1 - 2 | 110 | 62 | ||
| AK159793.1 | - | 1 - 2 | 73 | 62 | ||
| AK169654.1 | - | 1 - 2 | 84 | 62 | ||
| BC018291.1 | - | 1 - 2 | 63 | 4190167 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ddx56
Genomic Map