Assay Details
| Gene Symbol: Npc1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207242.2 | NP_997125.2 | 9 - 10 | 2672 | 98 | |
| GenBank mRNA | AK078947.1 | - | 9 - 10 | 2573 | 98 | |
| AY437866.1 | - | 9 - 10 | 2550 | 98 | ||
| BC131788.1 | - | 9 - 10 | 2549 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Npc1l1
Genomic Map