Assay Details
| Gene Symbol: Ddc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190448.1 | NP_001177377.1 | 11 - 12 | 1183 | 110 | |
| NM_016672.4 | NP_057881.1 | 11 - 12 | 1121 | 110 | ||
| GenBank mRNA | AF071068.1 | - | 11 - 12 | 1054 | 110 | |
| AK149141.1 | - | 11 - 12 | 1115 | 110 | ||
| BC060989.1 | - | 11 - 12 | 1139 | 110 | ||
| BC089594.1 | - | 12 - 13 | 1124 | 110 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ddc
Genomic Map