Assay Details
| Gene Symbol: Pabpc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130881.2 | NP_570951.2 | 4 - 5 | 1349 | 75 | |
| NM_148917.2 | NP_683717.1 | 4 - 5 | 975 | 75 | ||
| GenBank mRNA | AK135962.1 | - | 814 | 75 | ||
| BC003283.1 | - | 4 - 5 | 896 | 3582181 | 75 | |
| BC010345.1 | - | 4 - 5 | 975 | 3707766 | 75 | |
| BC056432.1 | - | 4 - 5 | 1349 | 5696369 | 75 | |
| BC058263.1 | - | 4 - 5 | 1561 | 75 | ||
| Gene Symbol: Gm5088 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002862.3 | 1 | 75 | |||
| GenBank mRNA | AK048750.1 | - | 1 | 75 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pabpc4
Genomic Map