Assay Details
| Gene Symbol: Mfsd2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029662.2 | NP_083938.2 | 7 - 8 | 967 | 88 | |
| GenBank mRNA | AK006096.1 | - | 7 - 8 | 966 | 88 | |
| BC046793.1 | - | 7 - 8 | 949 | 5717701 | 88 | |
| BC060526.1 | - | 7 - 8 | 938 | 5707105 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mfsd2a
Genomic Map