Assay Details
| Gene Symbol: Mfsd2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029662.2 | NP_083938.2 | 8 - 9 | 1092 | 71 | |
| GenBank mRNA | AK006096.1 | - | 8 - 9 | 1091 | 71 | |
| BC046793.1 | - | 8 - 9 | 1074 | 5717701 | 71 | |
| BC060526.1 | - | 8 - 9 | 1063 | 5707105 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mfsd2a
Genomic Map