Assay Details
| Gene Symbol: Nt5c1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085502.1 | NP_001078971.1 | 4 - 5 | 551 | 90 | |
| GenBank mRNA | BC147308.1 | - | 4 - 5 | 600 | 9056382 | 90 |
| BC147309.1 | - | 4 - 5 | 600 | 9056378 | 90 | |
| BC147730.1 | - | 4 - 5 | 595 | 9056679 | 90 | |
| BC147733.1 | - | 4 - 5 | 595 | 9056682 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nt5c1a
Genomic Map