Assay Details
| Gene Symbol: Ccdc23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038998.2 | NP_001034087.1 | 1 - 2 | 250 | 67 | |
| GenBank mRNA | CF105092.1 | - | 1 - 2 | 236 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc23
Genomic Map