Assay Details
| Gene Symbol: Rfx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159389.1 | NP_001152861.1 | 3 - 4 | 553 | 120 | |
| GenBank mRNA | AK049380.1 | - | 3 - 4 | 533 | 120 | |
| FJ804414.1 | - | 3 - 4 | 503 | 120 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rfx6
Genomic Map