Assay Details
| Gene Symbol: Mycbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019660.3 | NP_062634.2 | 1 - 2 | 77 | 67 | |
| GenBank mRNA | AB015858.1 | - | 1 - 2 | 40 | 67 | |
| AK033289.1 | - | 1 - 2 | 177 | 67 | ||
| AK037661.1 | - | 87 | 67 | |||
| AK077633.1 | - | 1 - 2 | 96 | 67 | ||
| AK077762.1 | - | 1 - 2 | 81 | 67 | ||
| AK077995.1 | - | 1 - 2 | 81 | 67 | ||
| AK145773.1 | - | 1 - 2 | 78 | 67 | ||
| BC028344.1 | - | 1 - 2 | 75 | 3985166 | 67 | |
| BC048478.1 | - | 1 - 2 | 258 | 6774657 | 67 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AB015858.1, AK037661.1, AK077633.1, AK077762.1, AK077995.1, AK145773.1, BC028344.1, BC048478.1, NM_019660.3 transcripts.
Set Membership: |
|

Mycbp
Genomic Map