Assay Details
| Gene Symbol: Mycbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019660.3 | NP_062634.2 | 3 - 4 | 197 | 91 | |
| GenBank mRNA | AB015858.1 | - | 3 - 4 | 160 | 91 | |
| AK037661.1 | - | 207 | 91 | |||
| AK077633.1 | - | 3 - 4 | 216 | 91 | ||
| AK077762.1 | - | 3 - 4 | 201 | 91 | ||
| AK077995.1 | - | 3 - 4 | 201 | 91 | ||
| AK132198.1 | - | 62 | 91 | |||
| AK145773.1 | - | 3 - 4 | 198 | 91 | ||
| BC028344.1 | - | 3 - 4 | 195 | 3985166 | 91 | |
| BC048478.1 | - | 3 - 4 | 378 | 6774657 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mycbp
Genomic Map