| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400010578.mm.1 |
NM_183163.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400010578.mm.1 |
NM_183163.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400010578.mm.1 |
TC0400001405.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400010576.mm.1 |
NM_183163.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400010576.mm.1 |
NM_183163.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400010576.mm.1 |
TC0400001405.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400010579.mm.1 |
NM_183163.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400010579.mm.1 |
NM_183163.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400010579.mm.1 |
TC0400001405.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400010582.mm.1 |
NM_183163.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400010582.mm.1 |
NM_183163.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400010582.mm.1 |
TC0400001405.mm.1 |
MTA TC |