Assay Details
| Gene Symbol: Rhbdl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183163.2 | NP_898986.2 | 3 - 4 | 508 | 69 | |
| GenBank mRNA | BX511550.1 | - | 2 - 3 | 286 | 69 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rhbdl2
Genomic Map