Assay Details
| Gene Symbol: Rhbdl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183163.2 | NP_898986.2 | 7 - 8 | 87 | ||
| GenBank mRNA | BC052650.1 | - | 7 - 8 | 87 | ||
| BM216905.1 | - | 1 - 2 | 370 | 87 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rhbdl2
Genomic Map