Assay Details
| Gene Symbol: Cyp4a14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007822.2 | NP_031848.1 | 1 - 2 | 223 | 89 | |
| GenBank mRNA | AK098088.1 | - | 1 - 2 | 220 | 89 | |
| AK149392.1 | - | 1 - 2 | 221 | 89 | ||
| AK165452.1 | - | 1 - 2 | 221 | 89 | ||
| BC089609.1 | - | 1 - 2 | 223 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cyp4a14
Genomic Map