Assay Details
| Gene Symbol: CK137956 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134733.1 | NP_001128205.1 | 2 - 3 | 1014 | 72 | |
| GenBank mRNA | BC147529.1 | - | 2 - 3 | 1014 | 9087439 | 72 |
More Information
Updated Mapping Information
Set Membership: |
|

CK137956
Genomic Map