Assay Details
| Gene Symbol: CK137956 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134733.1 | NP_001128205.1 | 4 - 5 | 1446 | 76 | |
| GenBank mRNA | BC147529.1 | - | 4 - 5 | 1446 | 9087439 | 76 |
More Information
Set Membership: |
|

CK137956
Genomic Map