Assay Details
| Gene Symbol: Stx7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB019212.1 | - | 1 - 2 | 82 | 72 | |
| AK002867.1 | - | 1 - 2 | 81 | 72 | ||
| AK013869.1 | - | 1 - 2 | 67 | 72 | ||
| AK082927.1 | - | 1 - 2 | 52 | 72 | ||
| AK142742.1 | - | 59 | 72 | |||
| AK166822.1 | - | 1 - 2 | 82 | 72 | ||
| AK169822.1 | - | 1 - 2 | 85 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB019212.1, AK002867.1, AK082927.1, AK142742.1, AK166822.1, AK169822.1 transcripts.
Set Membership: |
|

Stx7
Genomic Map