Assay Details
| Gene Symbol: Stx7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016797.4 | NP_058077.2 | 3 - 4 | 629 | 74 | |
| GenBank mRNA | AB019212.1 | - | 3 - 4 | 299 | 74 | |
| AK002867.1 | - | 3 - 4 | 298 | 74 | ||
| AK013869.1 | - | 3 - 4 | 283 | 74 | ||
| AK082927.1 | - | 3 - 4 | 269 | 74 | ||
| AK142742.1 | - | 276 | 74 | |||
| AK158923.1 | - | 3 - 4 | 630 | 74 | ||
| AK166822.1 | - | 3 - 4 | 299 | 74 | ||
| AK169822.1 | - | 3 - 4 | 302 | 74 | ||
| BC132123.1 | - | 2 - 3 | 179 | 40130400 | 74 | |
| BC132125.1 | - | 2 - 3 | 179 | 40130402 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Stx7
Genomic Map