Assay Details
| Gene Symbol: Stx7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016797.4 | NP_058077.2 | 6 - 7 | 912 | 84 | |
| GenBank mRNA | AB019212.1 | - | 6 - 7 | 582 | 84 | |
| AF056323.1 | - | 6 - 7 | 524 | 84 | ||
| AK002867.1 | - | 6 - 7 | 581 | 84 | ||
| AK013869.1 | - | 6 - 7 | 566 | 84 | ||
| AK082927.1 | - | 6 - 7 | 552 | 84 | ||
| AK142742.1 | - | 559 | 84 | |||
| AK158923.1 | - | 6 - 7 | 913 | 84 | ||
| AK166822.1 | - | 6 - 7 | 582 | 84 | ||
| AK169822.1 | - | 6 - 7 | 585 | 84 | ||
| AK198507.1 | - | 2 - 3 | 203 | 84 | ||
| AK213496.1 | - | 2 - 3 | 173 | 84 | ||
| BC132123.1 | - | 5 - 6 | 462 | 40130400 | 84 | |
| BC132125.1 | - | 5 - 6 | 462 | 40130402 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Stx7
Genomic Map