Assay Details
| Gene Symbol: Stx7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016797.4 | NP_058077.2 | 7 - 8 | 1013 | 157 | |
| GenBank mRNA | AB019212.1 | - | 7 - 8 | 683 | 157 | |
| AF056323.1 | - | 7 - 8 | 625 | 157 | ||
| AK002867.1 | - | 7 - 8 | 682 | 157 | ||
| AK013869.1 | - | 7 - 8 | 667 | 157 | ||
| AK082927.1 | - | 7 - 8 | 653 | 157 | ||
| AK142742.1 | - | 660 | 157 | |||
| AK158923.1 | - | 7 - 8 | 1014 | 157 | ||
| AK166822.1 | - | 7 - 8 | 683 | 157 | ||
| AK169822.1 | - | 7 - 8 | 686 | 157 | ||
| AK213496.1 | - | 3 - 4 | 274 | 157 | ||
| BC132123.1 | - | 6 - 7 | 563 | 40130400 | 157 | |
| BC132125.1 | - | 6 - 7 | 563 | 40130402 | 157 | |
More Information
Updated Mapping Information
Set Membership: |
|

Stx7
Genomic Map