Assay Details
| Gene Symbol: Tbc1d32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033385.3 | NP_001028557.2 | 19 - 20 | 2296 | 110 | |
| GenBank mRNA | AK035346.1 | - | 1 - 2 | 76 | 110 | |
| AK141206.1 | - | 4 - 5 | 491 | 110 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tbc1d32
Genomic Map