Assay Details
| Gene Symbol: Tbc1d32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033385.3 | NP_001028557.2 | 21 - 22 | 2529 | 116 | |
| GenBank mRNA | AK035346.1 | - | 3 - 4 | 309 | 116 | |
| AK141206.1 | - | 6 - 7 | 724 | 116 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tbc1d32
Genomic Map