Assay Details
| Gene Symbol: Tbc1d32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033385.3 | NP_001028557.2 | 23 - 24 | 2740 | 159 | |
| GenBank mRNA | AK035346.1 | - | 5 - 6 | 520 | 159 | |
| AK141206.1 | - | 8 - 9 | 935 | 159 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tbc1d32
Genomic Map