Assay Details
| Gene Symbol: Sun3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290519.1 | NP_001277448.1 | 1 - 2 | 37 | 112 | |
| GenBank mRNA | AK132922.1 | - | 1 - 2 | 38 | 112 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK132922.1, NM_001290519.1 transcripts.
Set Membership: |
|

Sun3
Genomic Map