Assay Details
| Gene Symbol: Speer5-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_001582.2 | 3 - 4 | 395 | 78 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | absent in melanoma 1 | Aim1 |
AI462491, AI463325 |
-47.1kb | Anti-sense | Chr. 10 - 43950307 - 44148924 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 40637 |
Gm40637 | 53.6kb | Anti-sense | Chr. 10 - 44267742 - 44268243 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Speer5-ps1
Genomic Map