Assay Details
| Gene Symbol: Heca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033432.3 | NP_001028604.1 | 2 - 3 | 1597 | 85 | |
| GenBank mRNA | AK132340.1 | - | 2 - 3 | 1598 | 85 | |
| AK134782.1 | - | 228 | 85 | |||
| BC094026.1 | - | 1 - 2 | 208 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Heca
Genomic Map