Assay Details
| Gene Symbol: Abracl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028440.1 | NP_082716.1 | 1 - 2 | 190 | 75 | |
| GenBank mRNA | AK171138.1 | - | 1 - 2 | 93 | 75 | |
| BC039801.1 | - | 1 - 2 | 69 | 75 | ||
| BC098509.1 | - | 1 - 2 | 132 | 75 | ||
| DV053786.1 | - | 1 - 2 | 239 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK171138.1, BC098509.1, NM_028440.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2127557 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2071271 | AK013984.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2071271 | DV053786.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2071271 | NM_028440.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 654158 | AK053473.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 831449 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 17079 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 398666 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 633236 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 882950 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 583733 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 700910 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 356490 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P108327 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P107425 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2071271 | AK013984.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2071271 | DV053786.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2071271 | NM_028440.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2127557 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P108327 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06989470 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P107425 | - | - |
| Whole Mouse Genome G4122A | A_51_P214679 | AK013984.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P214679 | AK171138.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P214679 | BC098509.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P214679 | DV053786.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P214679 | NM_028440.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P238873 | AK013984.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P238873 | AK171138.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P238873 | BC039801.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P238873 | BC098509.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P238873 | DV053786.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111146 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P108327 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P107425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26973077 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973023 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973101 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973117 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973139 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973105 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08801429 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973044 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973107 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973079 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08951721 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973062 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08295647 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973058 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973077 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973128 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973136 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973052 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973059 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973099 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08359214 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973029 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08742598 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973061 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973075 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973027 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973134 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973135 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973132 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973089 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973040 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973133 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973098 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973055 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973148 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08223738 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973072 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973155 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973109 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973076 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08742597 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973054 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973147 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973156 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973056 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973063 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973146 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973103 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973141 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973020 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973115 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08863648 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973119 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973082 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973113 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973071 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973086 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973070 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08295648 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973091 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973131 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973068 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973050 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973125 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973043 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973021 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973124 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973028 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973116 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08260286 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973087 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973048 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973092 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973144 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973022 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973152 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973129 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08242269 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973084 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08242279 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26973102 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10368043 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368046 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368044 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368042 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368039 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1426964_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10368030 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10368045 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10368041 | - | - |
| Mouse Genome 430 2.0 Array | 1426964_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1426964_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73112_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73112_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000013077.mm.1 | NM_028440.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000013077.mm.1 | NM_028440.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000013077.mm.1 | TC1000001802.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000013081.mm.1 | NM_028440.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000013081.mm.1 | NM_028440.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000013081.mm.1 | TC1000001802.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000013079.mm.1 | NM_028440.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000013079.mm.1 | NM_028440.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000013079.mm.1 | TC1000001802.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1426964_at | - | - |
| Mouse Expression Set 430 | 1426964_PM_at | - | - |
| Murine Genome U74v2 Set | 96135_at | - | - |
| Murine Genome U74v2 Set | 96135_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426964_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426964_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426964_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426964_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 487148 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4150402 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 105290215 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1257234 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1257234 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2546481 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 105290215 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 105290215 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2546481 | - | - |

Abracl
Genomic Map