Assay Details
| Gene Symbol: Reps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111065.1 | NP_001104535.1 | 10 - 11 | 1370 | 58 | |
| NM_009048.2 | NP_033074.2 | 11 - 12 | 1451 | 58 | ||
| GenBank mRNA | AF031939.1 | - | 10 - 11 | 1272 | 58 | |
| AK150284.1 | - | 11 - 12 | 1315 | 58 | ||
| AK151309.1 | - | 11 - 12 | 1315 | 58 | ||
| BC087547.1 | - | 10 - 11 | 1318 | 30441027 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Reps1
Genomic Map