Assay Details
| Gene Symbol: Reps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111065.1 | NP_001104535.1 | 7 - 8 | 999 | 106 | |
| NM_009048.2 | NP_033074.2 | 7 - 8 | 999 | 106 | ||
| GenBank mRNA | AF031939.1 | - | 6 - 7 | 820 | 106 | |
| AK041967.1 | - | 5 - 6 | 2077 | 106 | ||
| AK150284.1 | - | 7 - 8 | 863 | 106 | ||
| AK151309.1 | - | 7 - 8 | 863 | 106 | ||
| BC087547.1 | - | 7 - 8 | 947 | 30441027 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|

Reps1
Genomic Map