Assay Details
| Gene Symbol: Grhl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013756.1 | NP_001013778.1 | 7 - 8 | 1151 | 88 | |
| GenBank mRNA | BC089372.1 | - | 7 - 8 | 1151 | 30607679 | 88 |
More Information
Updated Mapping Information
Set Membership: |
|

Grhl3
Genomic Map