Assay Details
| Gene Symbol: Cpeb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290676.1 | NP_001277605.1 | 2 - 3 | 2550 | 90 | |
| NM_026252.4 | NP_080528.2 | 2 - 3 | 2550 | 90 | ||
| GenBank mRNA | AY313775.1 | - | 2 - 3 | 1280 | 90 | |
| BC079599.1 | - | 2 - 3 | 358 | 90 | ||
| BC145863.1 | - | 2 - 3 | 1274 | 40131166 | 90 | |
| BC145865.1 | - | 2 - 3 | 1274 | 40131168 | 90 | |
| CF182456.1 | - | 2 - 3 | 637 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cpeb4
Genomic Map