Assay Details
| Gene Symbol: Nprl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284359.1 | NP_001271288.1 | 12 - 13 | 1559 | 64 | |
| NM_001284360.1 | NP_001271289.1 | 10 - 11 | 1390 | 64 | ||
| NM_181569.3 | NP_853547.1 | 11 - 12 | 1520 | 64 | ||
| NR_104306.1 | 12 - 13 | 1740 | 64 | |||
| GenBank mRNA | AK036405.1 | - | 11 - 12 | 1429 | 64 | |
| AK039600.1 | - | 10 - 11 | 1366 | 64 | ||
| AK047190.1 | - | 12 - 13 | 1468 | 64 | ||
| AK081902.1 | - | 11 - 12 | 1431 | 64 | ||
| AK086378.1 | - | 1333 | 64 | |||
| AK090290.1 | - | 1300 | 64 | |||
| BC064471.1 | - | 11 - 12 | 1520 | 6334386 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nprl3
Genomic Map