Assay Details
| Gene Symbol: Srpk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019684.1 | NP_062658.1 | 8 - 9 | 797 | 83 | |
| GenBank mRNA | AF043288.1 | - | 8 - 9 | 797 | 83 | |
| AJ748651.1 | - | 7 - 8 | 621 | 83 | ||
| AK003681.1 | - | 2 - 3 | 159 | 83 | ||
| BC120883.1 | - | 8 - 9 | 792 | 40057934 | 83 | |
| BC120884.1 | - | 8 - 9 | 792 | 40057937 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Srpk3
Genomic Map