Assay Details
| Gene Symbol: Mecp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010788.4 | NP_034918.1 | 1 - 2 | 177 | 67 | |
| GenBank mRNA | AF158181.1 | - | 1 - 2 | 80 | 67 | |
| BC027153.1 | - | 1 - 2 | 99 | 4948925 | 67 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF158181.1, BC027153.1, NM_010788.4 transcripts.
Set Membership: |
|

Mecp2
Genomic Map