Assay Details
| Gene Symbol: Opn1mw | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008106.2 | NP_032132.1 | 1 - 2 | 135 | 77 | |
| GenBank mRNA | AF011389.1 | - | 1 - 2 | 121 | 77 | |
| AF190672.1 | - | 1 - 2 | 137 | 77 | ||
| AK149287.1 | - | 1 - 2 | 136 | 77 | ||
| BC014826.1 | - | 1 - 2 | 115 | 4507311 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Opn1mw
Genomic Map