Assay Details
| Gene Symbol: Slc6a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142809.1 | NP_001136281.1 | 2 - 3 | 1053 | 67 | |
| NM_001142810.1 | NP_001136282.1 | 2 - 3 | 1053 | 67 | ||
| NM_133987.2 | NP_598748.1 | 2 - 3 | 1053 | 67 | ||
| GenBank mRNA | AB077327.1 | - | 2 - 3 | 401 | 67 | |
| AF459435.1 | - | 2 - 3 | 1036 | 67 | ||
| AK180138.1 | - | 1 - 2 | 44 | 67 | ||
| AK214959.1 | - | 1 - 2 | 44 | 67 | ||
| BC024444.1 | - | 1 - 2 | 66 | 67 | ||
| BC024564.1 | - | 1 - 2 | 200 | 67 | ||
| BC049801.1 | - | 2 - 3 | 228 | 67 | ||
| BC141067.1 | - | 3 - 4 | 794 | 9055679 | 67 | |
| BC145319.1 | - | 3 - 4 | 458 | 9053874 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc6a8
Genomic Map