Assay Details
| Gene Symbol: L1cam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008478.3 | NP_032504.3 | 24 - 25 | 3409 | 78 | |
| GenBank mRNA | BC056988.1 | - | 24 - 25 | 3334 | 6822217 | 78 |
| X12875.1 | - | 23 - 24 | 3159 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

L1cam
Genomic Map