Assay Details
| Gene Symbol: Hdac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008229.2 | NP_032255.2 | 10 - 11 | 1299 | 95 | |
| GenBank mRNA | AB221663.1 | - | 10 - 11 | 1091 | 95 | |
| AK135572.1 | - | 5 - 6 | 605 | 95 | ||
| BC138517.1 | - | 10 - 11 | 1271 | 8861537 | 95 | |
| U31758.1 | - | 10 - 11 | 1259 | 95 | ||
More Information
Set Membership: |
|

Hdac2
Genomic Map