Assay Details
| Gene Symbol: Rfpl4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177783.1 | NP_001171254.1 | 2 - 3 | 889 | 103 | |
| GenBank mRNA | BG070936.2 | - | 1 - 2 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rfpl4b
Genomic Map