Assay Details
| Gene Symbol: Hivep2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010437.2 | NP_034567.2 | 6 - 7 | 6299 | 83 | |
| GenBank mRNA | AK137291.1 | - | 3 - 4 | 347 | 83 | |
| AK137384.1 | - | 2 - 3 | 289 | 83 | ||
| AK147244.1 | - | 7 - 8 | 6380 | 83 | ||
| AK147348.1 | - | 6 - 7 | 6298 | 83 | ||
| AK147419.1 | - | 11 - 12 | 6589 | 83 | ||
| AK147655.1 | - | 6 - 7 | 6298 | 83 | ||
| BC150704.1 | - | 3 - 4 | 5504 | 9087615 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hivep2
Genomic Map