Assay Details
| Gene Symbol: Atp6v0b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033617.3 | NP_291095.1 | 6 - 7 | 493 | 53 | |
| GenBank mRNA | AB060654.1 | - | 6 - 7 | 454 | 53 | |
| AF356006.1 | - | 6 - 7 | 497 | 53 | ||
| AK009495.1 | - | 6 - 7 | 484 | 53 | ||
| AK151365.1 | - | 6 - 7 | 464 | 53 | ||
| AK151847.1 | - | 6 - 7 | 481 | 53 | ||
| AK152330.1 | - | 6 - 7 | 498 | 53 | ||
| AK152823.1 | - | 6 - 7 | 481 | 53 | ||
| BC030393.1 | - | 6 - 7 | 478 | 4922226 | 53 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atp6v0b
Genomic Map