Assay Details
| Gene Symbol: Ebna1bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026932.4 | NP_081208.1 | 5 - 6 | 668 | 69 | |
| GenBank mRNA | AK007491.1 | - | 5 - 6 | 638 | 69 | |
| AK134732.1 | - | 5 - 6 | 623 | 69 | ||
| AK167070.1 | - | 5 - 6 | 623 | 69 | ||
| AK168269.1 | - | 5 - 6 | 595 | 69 | ||
| BC054723.1 | - | 5 - 6 | 651 | 5695176 | 69 | |
| BC062876.1 | - | 5 - 6 | 600 | 5686021 | 69 | |
More Information
Set Membership: |
|

Ebna1bp2
Genomic Map