Assay Details
| Gene Symbol: Cdc20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023223.2 | NP_075712.2 | 8 - 9 | 1431 | 77 | |
| GenBank mRNA | AB045313.1 | - | 8 - 9 | 1335 | 77 | |
| AF312208.1 | - | 8 - 9 | 1328 | 77 | ||
| AK011131.1 | - | 6 - 7 | 818 | 77 | ||
| AK011723.1 | - | 8 - 9 | 1430 | 77 | ||
| AK075998.1 | - | 8 - 9 | 1379 | 77 | ||
| AK083459.1 | - | 8 - 9 | 1260 | 77 | ||
| AK160204.1 | - | 8 - 9 | 1443 | 77 | ||
| AK168650.1 | - | 8 - 9 | 1383 | 77 | ||
| BC003215.1 | - | 8 - 9 | 1368 | 3157842 | 77 | |
More Information
Set Membership: |
|

Cdc20
Genomic Map