Assay Details
| Gene Symbol: Cfap57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026789.4 | NP_081065.2 | 20 - 21 | 3400 | 73 | |
| GenBank mRNA | AK003848.1 | - | 1 - 2 | 98 | 73 | |
| BC062805.1 | - | 1 - 2 | 105 | 1478343 | 73 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK003848.1, BC062805.1 transcripts.
Set Membership: |
|

Cfap57
Genomic Map