Assay Details
| Gene Symbol: Pes1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022889.3 | NP_075027.1 | 3 - 4 | 313 | 66 | |
| GenBank mRNA | AF289539.1 | - | 3 - 4 | 273 | 66 | |
| AK029075.1 | - | 3 - 4 | 313 | 66 | ||
| AK088831.1 | - | 3 - 4 | 291 | 66 | ||
| AK220142.1 | - | 3 - 4 | 260 | 66 | ||
| BC004844.1 | - | 3 - 4 | 298 | 2649598 | 66 | |
| BC011142.1 | - | 3 - 4 | 296 | 4207164 | 66 | |
More Information
Set Membership: |
|

Pes1
Genomic Map